Assay Details
| Gene Symbol: GHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556809.2 | XP_005556866.1 | 7 - 8 | 1829 | 154 | |
| XM_005556810.2 | XP_005556867.1 | 7 - 8 | 1389 | 154 | ||
| XM_005556811.2 | XP_005556868.1 | 7 - 8 | 1825 | 154 | ||
| XM_005556812.2 | XP_005556869.1 | 7 - 8 | 922 | 154 | ||
| XM_015451334.1 | XP_015306820.1 | 7 - 8 | 872 | 154 | ||
More Information
Set Membership: |
|

GHR
Genomic Map