Assay Details
| Gene Symbol: MBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587416.2 | XP_005587473.1 | 3 - 4 | 431 | 81 | |
| XM_005587417.1 | XP_005587474.1 | 3 - 4 | 335 | 81 | ||
| XM_015440694.1 | XP_015296180.1 | 3 - 4 | 745 | 81 | ||
| XM_015440695.1 | XP_015296181.1 | 3 - 4 | 1734 | 81 | ||
More Information
Set Membership: |
|

MBD3
Genomic Map