Assay Details
| Gene Symbol: SSC5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590404.2 | XP_005590461.1 | 4 - 5 | 639 | 133 | |
| XM_015441927.1 | XP_015297413.1 | 2 - 3 | 586 | 133 | ||
More Information
Set Membership: |
|

SSC5D
Genomic Map