Assay Details
| Gene Symbol: PAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556846.2 | XP_005556903.1 | 6 - 7 | 1237 | 104 | |
| XM_005556847.2 | XP_005556904.1 | 6 - 7 | 1066 | 104 | ||
| XM_005556848.2 | XP_005556905.1 | 6 - 7 | 1065 | 104 | ||
| XM_005556849.2 | XP_005556906.1 | 6 - 7 | 842 | 104 | ||
| XM_015451349.1 | XP_015306835.1 | 6 - 7 | 883 | 104 | ||
More Information
Set Membership: |
|

PAIP1
Genomic Map