Assay Details
| Gene Symbol: LOC102139497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005573990.2 | XP_005574047.1 | 4 - 5 | 1053 | 133 | |
| XM_015432584.1 | XP_015288070.1 | 4 - 5 | 763 | 133 | ||
| XM_015432585.1 | XP_015288071.1 | 5 - 6 | 663 | 133 | ||
| XM_015432586.1 | XP_015288072.1 | 5 - 6 | 748 | 133 | ||
| XM_015432588.1 | XP_015288074.1 | 5 - 6 | 817 | 133 | ||
More Information
Set Membership: |
|

LOC102139497
Genomic Map