Assay Details
| Gene Symbol: CD3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287688.1 | NP_001274617.1 | 2 - 3 | 279 | 108 | |
| XM_015435354.1 | XP_015290840.1 | 2 - 3 | 1099 | 108 | ||
| XM_015435355.1 | XP_015290841.1 | 2 - 3 | 426 | 108 | ||
| GenBank mRNA | AB073991.1 | - | 2 - 3 | 279 | 108 | |
More Information
Set Membership: |
|

CD3D
Genomic Map