Assay Details
| Gene Symbol: RNF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572442.2 | XP_005572499.1 | 3 - 4 | 406 | 147 | |
| XM_005572443.2 | XP_005572500.1 | 4 - 5 | 437 | 147 | ||
| XM_005572444.2 | XP_005572501.1 | 2 - 3 | 400 | 147 | ||
More Information
Set Membership: |
|

RNF34
Genomic Map