Assay Details
| Gene Symbol: NOC2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005545151.2 | XP_005545208.1 | 16 - 17 | 1994 | 90 | |
| XM_005545152.2 | XP_005545209.1 | 16 - 17 | 1994 | 90 | ||
More Information
Set Membership: |
|

NOC2L
Genomic Map