Assay Details
| Gene Symbol: ISG20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560453.2 | XP_005560510.1 | 3 - 4 | 795 | 59 | |
| XM_005560454.2 | XP_005560511.1 | 3 - 4 | 562 | 59 | ||
| XM_015453365.1 | XP_015308851.1 | 3 - 4 | 735 | 59 | ||
More Information
Set Membership: |
|

ISG20
Genomic Map