Assay Details
| Gene Symbol: TPPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285278.1 | NP_001272207.1 | 2 - 3 | 330 | 84 | |
| XM_005560742.1 | XP_005560799.1 | 2 - 3 | 363 | 84 | ||
| XM_015453234.1 | XP_015308720.1 | 2 - 3 | 366 | 84 | ||
| GenBank mRNA | AB179366.1 | - | 2 - 3 | 330 | 84 | |
More Information
Set Membership: |
|

TPPP2
Genomic Map