Assay Details
| Gene Symbol: AZI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005545576.2 | XP_005545633.1 | 4 - 5 | 633 | 87 | |
| XM_005545577.2 | XP_005545634.1 | 4 - 5 | 980 | 87 | ||
| XM_005545578.2 | XP_005545635.1 | 5 - 6 | 1065 | 87 | ||
| GenBank mRNA | AB168685.1 | - | 4 - 5 | 720 | 87 | |
| AB179144.1 | - | 4 - 5 | 540 | 87 | ||
More Information
Set Membership: |
|

AZI2
Genomic Map