Assay Details
| Gene Symbol: CPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285162.1 | NP_001272091.1 | 7 - 8 | 1354 | 105 | |
| GenBank mRNA | AB169871.1 | - | 7 - 8 | 1354 | 105 | |
More Information
Set Membership: |
|

CPE
Genomic Map