Assay Details
| Gene Symbol: LOC102133005 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549423.2 | XP_005549480.1 | 15 - 16 | 2153 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102133005| Gene Symbol: LOC102133005 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549423.2 | XP_005549480.1 | 15 - 16 | 2153 | 71 | |
Set Membership: |
|