Assay Details
| Gene Symbol: CTSO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556153.2 | XP_005556210.1 | 7 - 8 | 993 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTSO| Gene Symbol: CTSO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556153.2 | XP_005556210.1 | 7 - 8 | 993 | 138 | |
Set Membership: |
|