Assay Details
Gene Symbol: CD81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005576901.1 | XP_005576958.1 | 1 - 2 | 125 | 64 | |
XM_005576902.1 | XP_005576959.1 | 1 - 2 | 125 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
