Assay Details
| Gene Symbol: DLD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319401.1 | NP_001306330.1 | 3 - 4 | 283 | 89 | |
| XM_005550508.2 | XP_005550565.1 | 3 - 4 | 469 | 89 | ||
| GenBank mRNA | AB125164.1 | - | 3 - 4 | 287 | 89 | |
| AB168328.1 | - | 3 - 4 | 283 | 89 | ||
More Information
Set Membership: |
|

DLD
Genomic Map