Assay Details
| Gene Symbol: PSMC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284828.1 | NP_001271757.1 | 3 - 4 | 503 | 95 | |
| GenBank mRNA | AB168803.1 | - | 3 - 4 | 503 | 95 | |
More Information
Set Membership: |
|

PSMC4
Genomic Map