Assay Details
| Gene Symbol: PSMA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542477.2 | XP_005542534.1 | 8 - 9 | 765 | 120 | |
| XM_015431672.1 | XP_015287158.1 | 7 - 8 | 684 | 120 | ||
| GenBank mRNA | AB169279.1 | - | 1931 | 120 | ||
More Information
Set Membership: |
|

PSMA5
Genomic Map