Assay Details
| Gene Symbol: CWC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579395.2 | XP_005579452.1 | 2 - 3 | 249 | 126 | |
| XM_005579396.2 | XP_005579453.1 | 2 - 3 | 233 | 126 | ||
More Information
Set Membership: |
|

CWC15
Genomic Map