Assay Details
| Gene Symbol: HTATIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578395.2 | XP_005578452.1 | 2 - 3 | 372 | 83 | |
| XM_005578396.2 | XP_005578453.1 | 2 - 3 | 294 | 83 | ||
| XM_005578397.2 | XP_005578454.1 | 2 - 3 | 256 | 83 | ||
More Information
Set Membership: |
|

HTATIP2
Genomic Map