Assay Details
| Gene Symbol: FAM111A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577774.2 | XP_005577831.1 | 4 | 709 | 171 | |
| XM_005577775.2 | XP_005577832.1 | 3 | 823 | 171 | ||
| XM_005577776.2 | XP_005577833.1 | 3 | 257 | 171 | ||
| XM_015434677.1 | XP_015290163.1 | 5 | 933 | 171 | ||
| XM_015434678.1 | XP_015290164.1 | 5 | 894 | 171 | ||
| XM_015434679.1 | XP_015290165.1 | 4 | 748 | 171 | ||
| XM_015434680.1 | XP_015290166.1 | 3 | 269 | 171 | ||
More Information
Set Membership: |
|

FAM111A
Genomic Map