Assay Details
| Gene Symbol: FGG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556123.2 | XP_005556180.1 | 6 - 7 | 865 | 100 | |
| XM_005556124.2 | XP_005556181.1 | 6 - 7 | 865 | 100 | ||
More Information
Set Membership: |
|

FGG
Genomic Map