Assay Details
| Gene Symbol: SERPINA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562127.2 | XP_005562184.1 | 2 - 3 | 762 | 96 | |
| XM_005562128.2 | XP_005562185.1 | 2 - 3 | 751 | 96 | ||
More Information
Set Membership: |
|

SERPINA3
Genomic Map