Assay Details
| Gene Symbol: NR1H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571990.2 | XP_005572047.1 | 3 - 4 | 552 | 75 | |
| XM_005571991.2 | XP_005572048.1 | 3 - 4 | 552 | 75 | ||
More Information
Set Membership: |
|

NR1H4
Genomic Map