Assay Details
| Gene Symbol: DCTD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556368.2 | XP_005556425.1 | 3 - 4 | 605 | 107 | |
| XM_005556369.2 | XP_005556426.1 | 3 - 4 | 283 | 107 | ||
| XM_005556370.2 | XP_005556427.1 | 3 - 4 | 278 | 107 | ||
More Information
Set Membership: |
|

DCTD
Genomic Map