Assay Details
| Gene Symbol: TRIM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546255.2 | XP_005546312.1 | 1 - 2 | 231 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005546255.2 transcripts.
Set Membership: |
|

TRIM59
Genomic Map