Assay Details
| Gene Symbol: FUCA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552004.2 | XP_005552061.1 | 4 - 5 | 1358 | 105 | |
| XM_005552005.2 | XP_005552062.1 | 4 - 5 | 1358 | 105 | ||
| XM_015448736.1 | XP_015304222.1 | 4 - 5 | 953 | 105 | ||
More Information
Set Membership: |
|

FUCA2
Genomic Map