Assay Details
| Gene Symbol: ZFAND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563630.2 | XP_005563687.1 | 6 - 7 | 542 | 85 | |
| XM_005563632.2 | XP_005563689.1 | 6 - 7 | 496 | 85 | ||
| XM_005563634.2 | XP_005563691.1 | 6 - 7 | 475 | 85 | ||
More Information
Set Membership: |
|

ZFAND1
Genomic Map