Assay Details
| Gene Symbol: TP53BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540942.2 | XP_005540999.1 | 17 - 18 | 3645 | 93 | |
| XM_005540943.2 | XP_005541000.1 | 16 - 17 | 3470 | 93 | ||
| GenBank mRNA | AB173900.1 | - | 6 - 7 | 1541 | 93 | |
| AB174658.1 | - | 10 - 11 | 1765 | 93 | ||
More Information
Set Membership: |
|

TP53BP2
Genomic Map