Assay Details
| Gene Symbol: GNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552964.2 | XP_005553021.1 | 5 - 6 | 779 | 100 | |
| XM_015449395.1 | XP_015304881.1 | 4 - 5 | 983 | 100 | ||
More Information
Set Membership: |
|

GNMT
Genomic Map