Assay Details
| Gene Symbol: CASP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579488.2 | XP_005579545.1 | 4 - 5 | 927 | 75 | |
| XM_015435647.1 | XP_015291133.1 | 4 - 5 | 593 | 75 | ||
| GenBank mRNA | AB220461.1 | - | 4 - 5 | 477 | 75 | |
More Information
Set Membership: |
|

CASP1
Genomic Map