Assay Details
| Gene Symbol: FSCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550680.2 | XP_005550737.1 | 4 - 5 | 1280 | 94 | |
| XM_005550681.1 | XP_005550738.1 | 4 - 5 | 1323 | 94 | ||
More Information
Set Membership: |
|

FSCN3
Genomic Map