Assay Details
| Gene Symbol: LYRM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554124.2 | XP_005554181.1 | 1 - 2 | 308 | 133 | |
| XM_005554126.2 | XP_005554183.1 | 1 - 2 | 300 | 133 | ||
| XM_015449931.1 | XP_015305417.1 | 1 - 2 | 309 | 133 | ||
More Information
Set Membership: |
|

LYRM4
Genomic Map