Assay Details
| Gene Symbol: NMRAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591122.2 | XP_005591179.1 | 3 - 4 | 650 | 72 | |
| XM_005591123.2 | XP_005591180.1 | 3 - 4 | 534 | 72 | ||
| XM_015442415.1 | XP_015297901.1 | 4 - 5 | 648 | 72 | ||
More Information
Set Membership: |
|

NMRAL1
Genomic Map