Assay Details
| Gene Symbol: PLA2G1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572395.2 | XP_005572452.1 | 1 - 2 | 157 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005572395.2 transcripts.
Set Membership: |
|

PLA2G1B
Genomic Map