Assay Details
| Gene Symbol: SLC35B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554078.2 | XP_005554135.1 | 3 - 4 | 456 | 130 | |
| XM_005554079.2 | XP_005554136.1 | 3 - 4 | 551 | 130 | ||
| XM_005554080.2 | XP_005554137.1 | 3 - 4 | 820 | 130 | ||
| XM_005554081.2 | XP_005554138.1 | 3 - 4 | 539 | 130 | ||
| XM_005554082.2 | XP_005554139.1 | 3 - 4 | 456 | 130 | ||
| XM_015449911.1 | XP_015305397.1 | 2 - 3 | 406 | 130 | ||
| XM_015449912.1 | XP_015305398.1 | 2 - 3 | 491 | 130 | ||
| XM_015449913.1 | XP_015305399.1 | 2 - 3 | 395 | 130 | ||
More Information
Set Membership: |
|

SLC35B3
Genomic Map