Assay Details
| Gene Symbol: NIFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572894.2 | XP_005572951.1 | 4 - 5 | 787 | 141 | |
| GenBank mRNA | AB173165.1 | - | 5 - 6 | 807 | 141 | |
More Information
Set Membership: |
|

NIFK
Genomic Map