Assay Details
| Gene Symbol: RAB27A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559613.2 | XP_005559670.1 | 3 - 4 | 426 | 78 | |
| XM_005559614.2 | XP_005559671.1 | 4 - 5 | 506 | 78 | ||
| XM_005559615.2 | XP_005559672.1 | 2 - 3 | 1590 | 78 | ||
| XM_015452852.1 | XP_015308338.1 | 3 - 4 | 846 | 78 | ||
| XM_015452853.1 | XP_015308339.1 | 4 - 5 | 540 | 78 | ||
More Information
Set Membership: |
|

RAB27A
Genomic Map