Assay Details
| Gene Symbol: SLC66A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576553.2 | XP_005576610.1 | 6 - 7 | 688 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC66A3| Gene Symbol: SLC66A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576553.2 | XP_005576610.1 | 6 - 7 | 688 | 108 | |
Set Membership: |
|