Assay Details
| Gene Symbol: PREP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552253.2 | XP_005552310.2 | 5 - 6 | 1316 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PREP| Gene Symbol: PREP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552253.2 | XP_005552310.2 | 5 - 6 | 1316 | 83 | |
Set Membership: |
|