Assay Details
| Gene Symbol: ACTR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005571978.2 | XP_005572035.1 | 2 - 3 | 620 | 76 | |
| XM_005571979.2 | XP_005572036.1 | 2 - 3 | 621 | 76 | ||
| XM_005571980.2 | XP_005572037.1 | 2 - 3 | 236 | 76 | ||
More Information
Set Membership: |
|

ACTR6
Genomic Map