Assay Details
| Gene Symbol: RFC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572363.2 | XP_005572420.1 | 1 - 2 | 195 | 53 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RFC5| Gene Symbol: RFC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572363.2 | XP_005572420.1 | 1 - 2 | 195 | 53 | |
Set Membership: |
|