Assay Details
| Gene Symbol: UBE2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015444200.1 | XP_015299686.1 | 5 - 6 | 527 | 82 | |
| XM_015444201.1 | XP_015299687.1 | 4 - 5 | 434 | 82 | ||
More Information
Set Membership: |
|

UBE2A
Genomic Map