Assay Details
| Gene Symbol: IRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556392.2 | XP_005556449.1 | 2 - 3 | 296 | 68 | |
| XM_005556394.2 | XP_005556451.1 | 2 - 3 | 296 | 68 | ||
| XM_015451065.1 | XP_015306551.1 | 2 - 3 | 320 | 68 | ||
| XM_015451066.1 | XP_015306552.1 | 2 - 3 | 1609 | 68 | ||
More Information
Set Membership: |
|

IRF2
Genomic Map