Assay Details
| Gene Symbol: DYNC1LI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319409.1 | NP_001306338.1 | 4 - 5 | 545 | 87 | |
| GenBank mRNA | AB169497.1 | - | 4 - 5 | 545 | 87 | |
More Information
Set Membership: |
|

DYNC1LI2
Genomic Map