Assay Details
| Gene Symbol: TTI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005563036.1 | XP_005563093.1 | 3 - 4 | 1348 | 76 | |
| XM_015454662.1 | XP_015310148.1 | 3 - 4 | 1348 | 76 | ||
More Information
Set Membership: |
|

TTI2
Genomic Map