Assay Details
| Gene Symbol: PAK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589162.2 | XP_005589219.1 | 7 - 8 | 1683 | 84 | |
| XM_015441686.1 | XP_015297172.1 | 8 - 9 | 1756 | 84 | ||
| XM_015441687.1 | XP_015297173.1 | 8 - 9 | 1813 | 84 | ||
| XM_015441688.1 | XP_015297174.1 | 8 - 9 | 2108 | 84 | ||
| XM_015441689.1 | XP_015297175.1 | 8 - 9 | 1630 | 84 | ||
More Information
Set Membership: |
|

PAK4
Genomic Map