Assay Details
| Gene Symbol: SAA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001357502.1 | NP_001344431.1 | 3 - 4 | 324 | 123 | |
| GenBank mRNA | BB893422.1 | - | 3 - 4 | 381 | 123 | |
More Information
Set Membership: |
|

SAA4
Genomic Map