Assay Details
| Gene Symbol: PARP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547287.2 | XP_005547344.1 | 9 - 10 | 1670 | 88 | |
| XM_005547288.2 | XP_005547345.1 | 9 - 10 | 1665 | 88 | ||
| XM_005547289.2 | XP_005547346.1 | 9 - 10 | 1612 | 88 | ||
More Information
Set Membership: |
|

PARP3
Genomic Map