Assay Details
| Gene Symbol: RBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565974.2 | XP_005566031.1 | 5 - 6 | 681 | 109 | |
| XM_015456400.1 | XP_015311886.1 | 5 - 6 | 640 | 109 | ||
More Information
Set Membership: |
|

RBP4
Genomic Map